Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STOM Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | STOM |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-7672
![]() ![]() |
Proteintech Group Inc
120461AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Stomatin is an integral membrane protein found in lipid/protein-rich microdomains of almost all human tissues. It was named after the rare human disease haemolytic anaemia hereditary stomatocytosis. Stomatin is implicated in signal transduction and cell communication, and it may regulate cation movement through ion channels and transporters. Absence of Stomatin may cause Na+ and K+ ions to leak into and from erythrocytes. A second function of Stomatin may be to act as a cytoskeletal anchor. Stomatin is a major lipid-raft component of erythrocytes and epithelial cells, and is also an abundant platelet protein. It contains a single hydrophobic domain, close to the N-terminus, and a phosphorylation site.Specifications
STOM | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P27105 | |
2040 | |
STOM Fusion Protein Ag2676 | |
Primary | |
-20°C | |
STOM |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BND7, EPB7, EPB72, Protein 7.2b, STOM, stomatin | |
STOM | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title