Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STOX1 Antibody, Novus Biologicals™
SDP

Catalog No. NB395131 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395131 25 μL
NBP255177 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB395131 Supplier Novus Biologicals Supplier No. NBP25517725UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STOX1 Polyclonal specifically detects STOX1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen STOX1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias C10orf24, PEE4, storkhead box 1, storkhead-box protein 1, winged-helix domain-containing protein
Gene Symbols STOX1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DESRLMPASMTYLVSMESCAESAQENAAPISHCQSCQCFRDMHTQDVQEAPVAAEVTRKSHRGLGESVSWVQNGAVSVSAEHH
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219736
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.