Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STRA6 Antibody (496613), HRP, Novus Biologicals™
SDP

Catalog No. NB347193 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347193 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347193 Supplier Novus Biologicals Supplier No. FAB5904H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STRA6 Monoclonal antibody specifically detects STRA6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen STRA6
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 496613
Conjugate HRP
Dilution Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias FLJ12541, MCOPS9, stimulated by retinoic acid gene 6 homolog (mouse), stimulated by retinoic acid gene 6 protein homolog
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human STRA6, Met1-Pro667, Accession # AAH25256
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64220
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.