Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streck Temp-Chex™ Thermometer

Click to view available options
Temperature Range (Metric):
-30°C to +1°C
-5°C to +15°C
-90°C to +25°C
18°C to 50°C
For Use With (Application):
Freezers
Incubator
Refrigerator
Ultralow freezer
Thermometer Fill:
Red Spirit
Description
- Individually serial-numbered and NIST-traceable.
- Yellow background for easier reading.
- Enclosed in a durable plastic case to contain contents in case of breakage.
- A wide temperature range allows Temp-Chex thermometers to be used to monitor most laboratory applications.
- Mercury free and non-hazardous for increased laboratory safety.
- Thermometer probe is housed in a vial of propylene glycol, which creates a temperature buffer that prevents fluctuations.
Certifications
NIST, DKD Traceable: Each thermometer engraved with serial number traceable to NIST/DKD.
Specifications
Specifications
Certifications/Compliance | NIST, DKD |
For Use With (Application) | Freezers |
Includes | Temperature wall chart |
Thermometer Fill | Red Spirit |
Product Type | Thermometer |
Max. Temperature (Metric) | 1°C |
Min. Temperature (Metric) | -30°C |
Temperature Range (Metric) | -30°C to +1°C |
Accuracy | ±0.5°C |
Subdivision | 0.5° |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction