Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strep II Tag Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB219143 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB219143 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB219143 Supplier Novus Biologicals Supplier No. NBP241075H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Strep II Tag Polyclonal antibody specifically detects Strep II Tag in Epitope Tag samples. It is validated for Western Blot, ELISA

Specifications

Antigen Strep II Tag
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate HRP
Formulation PBS
Host Species Rabbit
Immunogen Antibody Strep II Tag antibody was raised against the amino acid sequence srWSHPQFEKsh.
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epitope Tags
Primary or Secondary Primary
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Target Molecule Epitope Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.