Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strep II Tag Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB344236 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344236 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344236 Supplier Novus Biologicals Supplier No. NBP241076PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Strep II Tag Polyclonal antibody specifically detects Strep II Tag in Epitope Tag samples. It is validated for Western Blot, ELISA

Specifications

Antigen Strep II Tag
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA
Formulation PBS
Host Species Rabbit
Immunogen Antibody Strep II Tag antibody was raised against the amino acid sequence asWSHPQFEKga.
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.