Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Strep-Tactin™ XT Sepharose™ Chromatography Resin

Catalog No. p-7184520
Click to view available options
Packaging Quantity:
10 mL
50 mL

Chromatography resin for purification of recombinant proteins tagged with a Strep-tag™ II and Twin-Strep-tag™.

  • Highly pure Strep-tag II and Twin-Strep-tag recombinant proteins
  • Fast purifications with simple elution
  • Physiological conditions and mild elution preserve target protein activity
  • Fast and easy regeneration
  • Compatible with a wide range of detergents, denaturants, and other additives
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Purification of recombinant proteins tagged with Strep-tag II and Twin-Strep-tag
Stability Stable in all commonly used aqueous buffers
Storage Requirements 2 to 8°C in 20% ethanol
Binding Capacity Approx. 10 mg protein with Strep-tag II or Twin-Strep-tag/mL of resin
Particle Size 34 μm
Matrix Rigid, cross-linked agarose, spherical
pH Range 6 to 10 pH
Density (Metric) Approx. 5 mg/mL resin
Ligand Type Strep-Tactin XT
Packaging Quantity 10 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.