Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptavidin Antibody (1C2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP104345 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP104345 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP104345 Supplier Novus Biologicals Supplier No. NBP104345

Mouse Monoclonal Antibody

Streptavidin Monoclonal specifically detects Streptavidin in Bacterial samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Streptavidin
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1C2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:3000, ELISA 1:100-1:2000
Gene Alias SA V1; SA V2; Streptavidin V1; Streptavidin V2
Host Species Mouse
Immunogen Recombinant Streptavidin (25-183aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Anti-Streptavidin mAb, clone 1C2, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant Streptavidin protein.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.