Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptavidin Antibody (1C2), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB310180 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310180 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310180 Supplier Novus Biologicals Supplier No. NBP104345MFV610

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Streptavidin Monoclonal antibody specifically detects Streptavidin in Bacteria samples. It is validated for Western Blot, ELISA

Specifications

Antigen Streptavidin
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1C2
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias SA V1; SA V2; Streptavidin V1; Streptavidin V2
Host Species Mouse
Immunogen Recombinant Streptavidin (25-183aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.