Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptococcus Agalactiae Antibody (224/46), HRP, Novus Biologicals™
SDP

Catalog No. NB10062933H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062933H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062933H Supplier Novus Biologicals Supplier No. NB10062933H

Mouse Monoclonal Antibody

Streptococcus Agalactiae Monoclonal specifically detects Streptococcus Agalactiae in Bacteria samples. It is validated for ELISA.

Specifications

Antigen Streptococcus Agalactiae
Applications ELISA
Classification Monoclonal
Clone 224/46
Conjugate HRP
Dilution ELISA
Formulation PBS with No Preservative
Gene Alias s agalactiae, S. agalactiae
Host Species Mouse
Immunogen Streptococcus agalactiae (NCTC 11360)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This product is specific for Streptococcus agalactiae a gram-positive streptococcus characterized by the presence of the group B Lancefield antigen.
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.