Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptococcus pneumoniae Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NBP229638 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229638 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP229638 Supplier Novus Biologicals Supplier No. NBP229638
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Streptococcus pneumoniae Polyclonal specifically detects Streptococcus pneumoniae in Bacteria samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Streptococcus pneumoniae
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS and BSA with 0.01% Sodium Azide
Gene Alias Pneumococcal surface protein A, PSPA
Host Species Rabbit
Immunogen Whole cell serotype blend
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-64502 recognizes Streptococcus pneumoniae and reacts with types 3, 4, 6, 7, 9, 14, 18, 19 and 23. S. pneumoniae is a non-motile gram-positive bacteria that is found in the human upper respiratory tract and often as diplococci (pairs of cocci). NB100-64502 is not absorbed and may react with related microorganisms.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.