Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptococcus pneumoniae CP5 Antibody (5F6) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB392761 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392761 0.025 mL
NBP274996 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB392761 Supplier Novus Biologicals Supplier No. NBP2749960.025ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Streptococcus pneumoniae CP5 Monoclonal specifically detects Streptococcus pneumoniae CP5 in Bacteria samples. It is validated for N/A.

Specifications

Antigen Streptococcus pneumoniae CP5
Classification Monoclonal
Clone 5F6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% Sodium Azide
Gene Alias Capsular Polysaccharide Serotype 5, CP5, Serotype 5 capsular polysaccharide
Host Species Mouse
Immunogen Streptococcus pneumoniae CP5
Purification Method Protein A purified
Quantity 0.025 mL
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity Streptococcus pneumoniae serotype 5 capsule
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.