Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streptococcus pneumoniae CP5 Antibody (5F6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $499.50
Specifications
Antigen | Streptococcus pneumoniae CP5 |
---|---|
Clone | 5F6 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Streptococcus pneumoniae CP5 Monoclonal specifically detects Streptococcus pneumoniae CP5 in Bacteria samples. It is validated for N/A.Specifications
Streptococcus pneumoniae CP5 | |
Monoclonal | |
Purified | |
Immunology, Virology Bacteria and Parasites | |
Streptococcus pneumoniae CP5 | |
Primary | |
Streptococcus pneumoniae serotype 5 capsule |
5F6 | |
Unconjugated | |
Mouse | |
Capsular Polysaccharide Serotype 5, CP5, Serotype 5 capsular polysaccharide | |
IgG3 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title