Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptococcus Pneumoniae Type 1 Antibody (1040/208), Novus Biologicals™
SDP

Catalog No. NB10064570 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064570 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064570 Supplier Novus Biologicals Supplier No. NB10064570
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Streptococcus Pneumoniae Type 1 Monoclonal specifically detects Streptococcus Pneumoniae Type 1 in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Streptococcus Pneumoniae Type 1
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1040/208
Concentration 2.7 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Host Species Mouse
Immunogen Native
Purification Method IgM purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-64570 recognizes Streptococcus pneumoniae, a causative agent of respiratory disease in humans
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.