Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptococcus streptokinase Antibody, Novus Biologicals™
SDP

Catalog No. NB017520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017520 0.1 mL
NB10062936 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017520 Supplier Novus Biologicals Supplier No. NB100629360.1ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Streptococcus streptokinase Polyclonal specifically detects Streptococcus streptokinase in Bacteria, Plant samples. It is validated for ELISA, Western Blot.

Specifications

Antigen Streptococcus streptokinase
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:20-1:200
Formulation Phosphate Buffered Saline with 0.09% Sodium Azide
Host Species Rabbit
Immunogen Streptococcus streptokinase bacterial protein prepared from Lancefield strain of Streptococcus-beta hemolyticus.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This product is specific for Streptococcus streptokinase, a metallo-enzyme produced by various strains of Streptococcus. It is 415 amino acids in length with a MW of approximately 47kDa. Streptokinase can be used as an anti thrombotic because it can liquify blood clots by converting plasminogen to plasmin.
Target Species Bacteria, Plant
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.