Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva StrepTrap™ HP Prepacked Columns

Catalog No. 45001520 Shop All Cytiva Products
missing translation for 'orderingAttributeHoverText'
Quantity:
1 x 5mL
Flow Rate:
<20 mL/min.
Content And Storage:
4° to 8°C

Purify Strep(II)-tagged recombinant proteins by affinity chromatography. Cytiva Life Sciences™ StrepTrap™ HP Prepacked Columns are packed with a robust and stable Sepharose™ based medium which results in high resolution, sharp concentrated peaks, and reproducible purifications. The binding capacity is also high.

  • Prepacked with StrepTactin Sepharose High Performance
  • Binding capacity Approx. 6 mg Strep II-tagged protein/ml medium
  • Maximum pressure: 3 bar
  • Column material: polypropylene
  • Compatible with a wide range of reducing agents, detergents, denaturants, and other additives
  • Generated with 0.5M NaOH
  • 1 and 5mL columns

Media
  • Ligand: Streptactin
  • Matrix: 6% cross-linked agarose
  • Average particle size: 34μm
  • pH stability working range: >7

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.

5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.

TRUSTED_SUSTAINABILITY

Specifications

Product Type StrepTactin Sepharose HP prepacked column
Bed Volume 1mL
Diameter (English) Inner 0.62 in.
Diameter (Metric) Inner 16mm
Quantity 1 x 5mL
Height (English) 0.98 in.
Height (Metric) 25mm
Flow Rate <20 mL/min.
Material Polypropylene
Ligand Type StrepTactin (∼5mg/mL medium)
Max. Pressure 3 bar
pH Range 7
Particle Size 34μm
Support Type 6% Highly Cross-linked Agarose
Content And Storage 4° to 8°C
Column Type Ion Exchange Column
For Use With (Application) Purification of Strep (II) -tagged recombinant Proteins by affinity chromato graphy
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.