Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Striated Muscle Myosin Heavy Chain Antibody (83B6), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB309699 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309699 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309699 Supplier Novus Biologicals Supplier No. NBP250254APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Striated Muscle Myosin Heavy Chain Monoclonal antibody specifically detects Striated Muscle Myosin Heavy Chain in Human, Mouse, Rat, Chicken, Rabbit samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen Striated Muscle Myosin Heavy Chain
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 83B6
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Host Species Mouse
Immunogen Purified human skeletal muscle myosin
Purification Method Ascites
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat, Chicken, Rabbit
Content And Storage Store at 4C in the dark.
Form Ascites
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.