Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Striated Muscle Myosin Heavy Chain Antibody (83B6), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NBP50254549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP50254549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP50254549 Supplier Novus Biologicals Supplier No. NBP250254JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Striated Muscle Myosin Heavy Chain Monoclonal specifically detects Striated Muscle Myosin Heavy Chain in Human, Mouse, Rat, Chicken, Rabbit samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Striated Muscle Myosin Heavy Chain
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 83B6
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Purified human skeletal muscle myosin
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cardiovascular Biology, Cellular Signaling
Primary or Secondary Primary
Test Specificity This antibody reacts with all skeletal and cardiac muscle type myosin heavy chains (striated muscle myosin heavy chains SM1 and SM2).
Target Species Chicken, Human, Mouse, Rabbit, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.