Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Striated Muscle Troponin I Antibody (42/25), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP20309A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20309A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20309A94 Supplier Novus Biologicals Supplier No. NBP250309AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Striated Muscle Troponin I Monoclonal specifically detects Striated Muscle Troponin I in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot.

Specifications

Antigen Striated Muscle Troponin I
Applications Western Blot
Classification Monoclonal
Clone 42/25
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Skeletal muscle troponin I
Host Species Mouse
Immunogen Purified rabbit skeletal muscle troponin I
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cellular Signaling
Primary or Secondary Primary
Test Specificity Specific for striated muscle troponin I.
Target Species Human, Mouse, Rat, Chicken
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.