Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Striated Muscle Troponin I Antibody (42/25), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB309607 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309607 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309607 Supplier Novus Biologicals Supplier No. NBP250309MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Striated Muscle Troponin I Monoclonal antibody specifically detects Striated Muscle Troponin I in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot

Specifications

Antigen Striated Muscle Troponin I
Applications Western Blot
Classification Monoclonal
Clone 42/25
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Skeletal muscle troponin I
Host Species Mouse
Immunogen Purified rabbit skeletal muscle troponin I
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling
Primary or Secondary Primary
Target Species Human, Mouse, Rat, Chicken
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.