Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STRN4 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB096159 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096159 100 μL
NB096160 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB096159 Supplier Novus Biologicals Supplier No. NBP304871100UL

Rabbit Polyclonal Antibody

STRN4 Polyclonal specifically detects STRN4 in Rat samples. It is validated for Western Blot.

Specifications

Antigen STRN4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias FLJ35594, striatin, calmodulin binding protein 4, striatin-4, ZIN, zinedin
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 230-320 of human STRN4 (NP_037535.2).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29888
Target Species Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.