Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium fluoride, Puratronic™, 99.99% (metals basis)

Catalog No. p-4782163
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1087806 5 g
AA1087814 25 g
AA1087822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1087806 Supplier Thermo Scientific Chemicals Supplier No. 01087806
Only null left
Add to Cart

CAS: 7783-48-4 | F2Sr | 125.62 g/mol

Strontium fluoride is used as an optical coating on lenses, and as a thermoluminescent dosimeter crystal. It also finds use as a water insoluble strontium source for use in oxygen-sensitive applications such as metal production. It serves both as a source of strontium and fluoride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-48-4
Molecular Formula F2Sr
Molecular Weight (g/mol) 125.62
MDL Number MFCD00011251
InChI Key FVRNDBHWWSPNOM-UHFFFAOYSA-L
Synonym strontium difluoride, unii-efy8gjs81z, strontium fluoride srf2, efy8gjs81z, strontium 2+ ion difluoride, fluoride, sr, fluoride, strontium 2+ difluoride, beta-bromo isovaleric acid, ksc379a4r, strontium fluoride 250g
PubChem CID 82210
SMILES [F-].[F-].[Sr++]

Specifications

Melting Point 1473°C
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.99%, metal basis
Solubility Information Insoluble in water.
IUPAC Name strontium(2+) difluoride
Formula Weight 125.62
Refractive Index 1.442
Density 4.24
Chemical Name or Material Strontium Fluoride
Hazard Category H315-H319-H335
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 232-000-3
RTECSNumber WK8925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.