Learn More
Strontium fluoride, 99% min
Description
Strontium fluoride is used as an optical material for a small range of special applications. It also used as use is as a carrier of strontium-90 radioisotope in radioisotope thermoelectric generators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1473°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 250 g |
| Merck Index | 14,8842 |
| Solubility Information | Slightly soluble in water (11;7mg at 18°C/100mL), dilute acids; Decomposes in strong acids |
| Formula Weight | 125.62 |
| Percent Purity | ≤99% |
| Odor | Odorless |
| Refractive Index | 1.442 |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 232-000-3 |
| RTECSNumber | WK8925000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.