Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium sulfate, Reagent Grade

Catalog No. aa3837314
Click to view available options
Quantity:
25 g
100 g

Catalog No. AA3837314

Supplier: Thermo Scientific Chemicals 03837314

Only null left
Add to Cart

CAS: 7759-02-6 | O4SSr | 183.68 g/mol

Strontium sulfate is used as a precursor to other strontium compounds and as ceramic precursor. Its application in nitrate for use in pyrotechnics. Used as a fining agent in crystal glass. It is also used in powder coating, liquid paints and plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7759-02-6
ChEBI CHEBI:63041

Specifications

Melting Point 1600°C
Physical Form Crystalline
Quantity 25 g
Molecular Formula Cl2Mn
MDL Number MFCD00011114
Merck Index 14,8848
Synonym manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline
Solubility Information Soluble in dilute hydrochloric acid and nitric acid . Slightly soluble in water < 0.01g/L . Insoluble in ethanol and alkalis.
InChI Key GLFNIEUTAYBVOC-UHFFFAOYSA-L
SMILES [Cl-].[Cl-].[Mn++]
IUPAC Name manganese(2+) dichloride
Molecular Weight (g/mol) 125.84
PubChem CID 10313134
Formula Weight 183.68
Grade Reagent
Density 3.960 g/mL
Chemical Name or Material Strontium sulfate, For reagent grade
Show More Show Less
EINECSNumber 231-850-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.