Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ STX 64
GREENER_CHOICE

Catalog No. p-7222994
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent steroid sulfatase inhibitor; also inhibits carbonic anhydrase II

STX 64 is a potent steroid sulfatase inhibitor (IC50 = 8 nM). Estrone sulfate-stimulated uterine growth and mammary tumor growth are inhibited by STX 64 in animal models. STX 64 shows inhibition of steroid sulfatase activity in peripheral blood lymphocytes, liver and tumor tissues and is active in vivo. STX 64 is also a potent inhibitor of carbonic anhydrase II (IC50 = 25 nM).

Specifications

Chemical Name or Material 6,7,8,9,10,11-Hexahydro-6-oxobenzo[b]cyclohepta[d]pyran-3-yl sulfamate
CAS 288628-05-7
Quantity 25 mg
Target Hydrolase Inhibitors
Molecular Formula C14H15NO5S
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.