Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STX8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551597
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551597 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551597 Supplier Invitrogen™ Supplier No. PA551597
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YDSTCQIAQE IAEKIQQRNQ YERKGEKAPK LTVTIRALLQ NLKEKIALLK DLLLRAVSTH QITQLEGDRR QNLLDDLVTR ERLLLASFKN EGAEPDLIRS SLMSEEAKRG APNPWLFEEP EETRGLGFDE IRQQQQKIIQ EQDAGLD Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.

The gene is a member of the syntaxin family. The encoded protein is involved in protein trafficking from early to late endosomes via vesicle fusion and exocytosis. A related pseudogene has been identified on chromosome 12. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STX8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STX8
Gene Accession No. Q9UNK0
Gene Alias 0610007H08Rik; 1110002H11Rik; 4930571E13Rik; CARB; CIP-1-associated regulator of cyclin B; Stx8; syntaxin 8; syntaxin-8; Syntaxin-like protein 3I35
Gene Symbols STX8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STX8. Recombinant protein control fragment (Product #RP-95932).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9482
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.