Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stxbp5l Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB10091288 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10091288 0.5 mg
NB017896 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10091288 Supplier Novus Biologicals Supplier No. NB10091288
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Stxbp5l Polyclonal specifically detects Stxbp5l in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Stxbp5l
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 10-50 ug/ml, Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Formulation Lyophilized from PBS with No Preservative
Gene Accession No. Q5DQR4
Gene Alias KIAA1006, lethal(2) giant larvae protein homolog 4, syntaxin binding protein 5-like
Gene Symbols STXBP5L
Host Species Rabbit
Immunogen A synthetic peptide from amino acid region 900-950 of mouse Stxbp5l conjugated to blue carrier protein was used as the antigen. The peptide is homologous in rat.
Purification Method Ammonium sulfate precipitation
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9515
Test Specificity Stxbp5l.
Reconstitution Reconstitute 0.5 mg size in 0.5 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the 0.1 mg size is provided in reconstituted format.
Target Species Mouse, Rat, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.