Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STYK1 Antibody (484713), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB376022 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376022 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376022 Supplier Novus Biologicals Supplier No. FAB6668L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STYK1 Monoclonal specifically detects STYK1 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Western Blot.

Specifications

Antigen STYK1
Applications CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 484713
Conjugate DyLight 550
Dilution Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias DKFZp761P1010, EC 2.7.10.2, NOKProtein PK-unique, serine/threonine/tyrosine kinase 1Novel oncogene with kinase domain, SuRTK106, tyrosine-protein kinase STYK1
Host Species Mouse
Immunogen E. coli-derived recombinant human STYK1, Asp72-Val320, Accession # Q6J9G0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 55359
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.