Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STYXL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | STYXL1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
STYXL1 Polyclonal specifically detects STYXL1 in Human samples. It is validated for Western Blot.Specifications
STYXL1 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
dual specificity phosphatase 24 (putative), Dual specificity phosphatase inhibitor MK-STYX, Dual specificity protein phosphatase 24, DUSP24, Map kinase phosphatase-like protein MK-STYX, MKSTYX, MK-STYX, serine/threonine/tyrosine interacting-like 1, serine/threonine/tyrosine-interacting-like protein 1 | |
STYXL1 | |
IgG | |
36 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_057170 | |
51657 | |
Synthetic peptide directed towards the middle region of human STYXL1The immunogen for this antibody is STYXL1. Peptide sequence FLRTQKIIWMPQELDAFQPYPIEIVPGKVFVGNFSQACDPKIQKDLKIKA. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title