Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STYXL1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220528 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220528 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220528 Supplier Novus Biologicals Supplier No. NBP220528
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STYXL1 Polyclonal specifically detects STYXL1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen STYXL1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9Y6J8
Gene Alias dual specificity phosphatase 24 (putative), Dual specificity phosphatase inhibitor MK-STYX, Dual specificity protein phosphatase 24, DUSP24, Map kinase phosphatase-like protein MK-STYX, MKSTYX, MK-STYX, serine/threonine/tyrosine interacting-like 1, serine/threonine/tyrosine-interacting-like protein 1
Gene Symbols STYXL1
Host Species Rabbit
Immunogen Full length human STYXL1 Recombinant protein.
Molecular Weight of Antigen 36 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51657
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.