Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SU 11274, Tocris Bioscience™

Catalog No. p-4816885
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective inhibitor of MET kinase activity

Selective inhibitor of MET tyrosine kinase activity (IC50 = 0.01 μM in vitro). Reduces cell growth in a dose-dependent manner; induces cell cycle arrest and apoptosis. Abrogates cell motility and migration in vitro and tumor angiogenesis in vivo.

Chemical Identifiers

CAS 658084-23-2
Molecular Formula C28H30ClN5O4S
Molecular Weight (g/mol) 568.089
InChI Key FPYJSJDOHRDAMT-KQWNVCNZSA-N
Synonym met kinase inhibitor, 3z-n-3-chlorophenyl-3-3,5-dimethyl-4-4-methylpiperazin-1-yl carbonyl-1h-pyrrol-2-yl methylene-n-methyl-2-oxo-2,3-dihydro-1h-indole-5-sulfonamide, 3z-n-3-chlorophenyl-3-3,5-dimethyl-4-4-methylpiperazine-1-carbonyl-1h-pyrrol-2-yl methylidene-n-methyl-2-oxo-1h-indole-5-sulfonamide, z-n-3-chlorophenyl-3-3,5-dimethyl-4-4-methylpiperazine-1-carbonyl-1h-pyrrol-2-yl methylene-n-methyl-2-oxoindoline-5-sulfonamide, pubchem19146, d0e1qi, su-mi-2, n-3-chlorophenyl-n-methyl-3-3,5-dimethyl-4-4-methylpiperazin-1-yl carbonyl-1h-pyrrol-2-yl methylene-2-oxo-2,3-dihydro-1h-indole-5-sulfonamide, su hplc , powder
PubChem CID 9549297
IUPAC Name (3Z)-N-(3-chlorophenyl)-3-[[3,5-dimethyl-4-(4-methylpiperazine-1-carbonyl)-1H-pyrrol-2-yl]methylidene]-N-methyl-2-oxo-1H-indole-5-sulfonamide
SMILES CC1=C(NC(=C1C(=O)N2CCN(CC2)C)C)C=C3C4=C(C=CC(=C4)S(=O)(=O)N(C)C5=CC(=CC=C5)Cl)NC3=O

Specifications

Quantity 10 mg
Formula Weight 568.09
Percent Purity >97%
Chemical Name or Material SU 11274
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.