Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SU 16f
GREENER_CHOICE

Catalog No. 330410
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective PDGFRβ inhibitor

Potent and selective platelet-derived growth factor receptor β (PDGFRβ) inhibitor (IC50 = 10 nM) that displays > 14-fold, > 229-fold and > 10000-fold selectivity over VEGFR2, FGFR1 and EGFR respectively. Inhibits proliferation of HUVEC and NIH3T3 cells in vitro (IC50 = 0.11 μM).

Chemical Identifiers

CAS 251356-45-3
Molecular Formula C24H22N2O3

Specifications

Quantity 10 mg
Formula Weight 386.44
Purity >98%
Molecular Weight (g/mol) 386.451
Chemical Name or Material SU 16f
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.