Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SU 5416
GREENER_CHOICE

Catalog No. p-4784609
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

VEGFR inhibitor. Also inhibits KIT, RET, MET and FLT3

Inhibitor of vascular endothelial growth factor receptor (VEGFR) that also inhibits other tyrosine kinases KIT, MET, FLT3 and RET. Displays no activity against EGFR, HER2, IGF1R and PDGFR. Inhibits tumor vascularization and growth of multiple tumor types.

Chemical Identifiers

CAS 204005-46-9
Molecular Formula C15H14N2O

Specifications

Quantity 10 mg
Formula Weight 238.28
Purity >99%
Molecular Weight (g/mol) 238.29
Chemical Name or Material SU 5416
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.