Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hoefer™ SUBHT High Throughput Submarine Gel Unit
SDP

Catalog No. p-4596929
Encompass
Click to view available options
Type:
Colored Loading Strip

Includes

  • Buffer chamber assembly
  • Safety lid, high-voltage leads
  • Fluorescent Labeled Running Tray with Casting Gates
  • Four 1mm thick 30-well combs
  • Colored Loading Strips
  • Two Buffer Recirculation Ports

Ideal for high throughput screening applications like single strand polymorphism (SSP) analysis following PCR

  • Fast, error-free loading: load an entire 96-well plate using a multichannel pipetter
  • Simplified Analysis: Fluorescent labels for each sample printed on a UV transparent gel tray allows for instant sample identification (matched to a 96-well plate) during post-run visualization
  • High Throughput: Run up to 120 samples (96-well plate plus 24 control and/or marker samples) in 30 minutes
  • Better Reproducibility: Eliminate variability caused by running buffer pH drift and ionic gradient formation with optional buffer recirculation capability
  • Reduced Buffer Needs: Compact tank design needs less buffer to cover gel which not only reduces buffer consumption but provides greater control of voltage gradients and run times
  • Maximum Flexibility: Fully compatible with a wide range of combs and accessories to meet all your high throughput and standard gel analysis needs
  • This high throughput horizontal gel unit can accommodate up to 120 samples and resolve DNA strands from 50-1000 base pairs in 30 minutes
Specifications
  • Gel Dimensions: 15 x 15 cm
  • Maximum Buffer Volume: 1.2L
  • Maximum Power Settings: 300V, 200mA, 60W
  • Maximum Sample Capacity: 120
  • Maximum Running Temperature: 45°C
  • Indoor Use: 4-40°C
  • Humidity: up to 80%
  • Unit Dimensions: 33.5L x 21.5W x 7 cmH
  • Safety Certifications: EN61010-1, CE

Applications:

Single Strand Polymorphism (SSP) analysis

Specifications

For Use With (Equipment) SUBHT High Throughput Submarine Gel Unit
Type Colored Loading Strip
Includes Buffer chamber assembly, safety lid with high voltage Leads, fluorescent labeled running tray with casting gates, four 1mm thick 30-well combs, colored loading strips, two Buffer recirculation ports
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.