Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Succinic Anhydride, 99%
Click to view available options
Quantity:
5 g
50 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 116.0°C to 120.0°C |
| Boiling Point | 261.0°C |
| Quantity | 5 g |
| Packaging | Glass Bottle |
| Assay Percent Range | 99% |
| Viscosity | 3 mPa.s (125°C) |
| Beilstein | 17,407 |
| Fieser | 04,468 |
| Merck Index | 15,9 |
| Solubility Information | Solubility in water: 62g/L (20°C). Other solubilities: soluble in chloroform,carbon tetrachloride and,alcohol,slightly soluble in ether,20g/L benzene (20°C) |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes severe skin burns and eye damage. May cause respiratory irritation. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled |
| Precautionary Statement | GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection. Immediately |
| EINECSNumber | 203-570- |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction