Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUCLA2 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB047401 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB047401 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB047401 Supplier Novus Biologicals Supplier No. NBP297755C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SUCLA2 Polyclonal specifically detects SUCLA2 in Human samples. It is validated for Western Blot.

Specifications

Antigen SUCLA2
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias A-BETA, ATP-specific succinyl-CoA synthetase subunit beta, beta subunit, EC 6.2.1, EC 6.2.1.5, mitochondrial succinyl-CoA ligase [ADP-forming] subunit beta, MTDPS5, Renal carcinoma antigen NY-REN-39, succinate-CoA ligase, ADP-forming, beta subunit, succinyl-CoA ligase [ADP-forming] subunit beta, mitochondrial
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SUCLA2 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 8803
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.