Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUCLG1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB085494 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB085494 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB085494 Supplier Novus Biologicals Supplier No. NBP299517AF532

Rabbit Polyclonal Antibody

SUCLG1 Polyclonal specifically detects SUCLG1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SUCLG1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias EC 6.2.1, EC 6.2.1.4, FLJ21114, FLJ43513, GALPHA, MTDPS9, SCS-alpha, succinate-CoA ligase, alpha subunit, succinate-CoA ligase, GDP-forming, alpha subunit, succinyl-CoA ligase [GDP-forming] subunit alpha, mitochondrial, Succinyl-CoA synthetase subunit alpha, SUCLA1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human SUCLG1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8802
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.