Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUCNR1/GPR91 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP100861X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
Conjugate:
Alexa Fluor 488
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
NBP100861X 0.1 mL Alexa Fluor 488
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP100861X Supplier Novus Biologicals Supplier No. NBP100861AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects SUCNR1/GPR91 in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blotting

SUCNR1/GPR91 Polyclonal specifically detects SUCNR1/GPR91 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Product Image.

Specifications

Antigen SUCNR1/GPR91
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q9BXA5
Gene Alias GPR91G protein-coupled receptor 91, G-protein coupled receptor 91, P2Y purinoceptor 1, P2Y purinoceptor 1-like, succinate receptor 1
Gene Symbols SUCNR1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human GPR91 protein (between residues 100-200) [Uniprot: Q9BXA5]
Molecular Weight of Antigen 38 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 56670
Test Specificity GPR91 (A135) detects endogenous levels of GPR91 protein.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.