Learn More
Description
Specifically detects SUCNR1/GPR91 in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blotting
Specifications
Specifications
| Antigen | SUCNR1/GPR91 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | PBS with No Preservative |
| Gene Accession No. | Q9BXA5 |
| Gene Alias | GPR91G protein-coupled receptor 91, G-protein coupled receptor 91, P2Y purinoceptor 1, P2Y purinoceptor 1-like, succinate receptor 1 |
| Gene Symbols | SUCNR1 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
