Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUCNR1/GPR91 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB338648 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338648 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338648 Supplier Novus Biologicals Supplier No. NBP100861MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SUCNR1/GPR91 Polyclonal antibody specifically detects SUCNR1/GPR91 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), In vitro Assay

Specifications

Antigen SUCNR1/GPR91
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), In vitro Assay
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, In vitro assay
Formulation 50mM Sodium Borate
Gene Alias GPR91G protein-coupled receptor 91, G-protein coupled receptor 91, P2Y purinoceptor 1, P2Y purinoceptor 1-like, succinate receptor 1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human GPR91 protein (between residues 100-200) [Uniprot: Q9BXA5]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 56670
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.