Learn More
Ammonium metavanadate, 99.996%, (trace metal basis)
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 200.0°C |
| Color | White to Yellow |
| Physical Form | Crystalline Powder |
| Quantity | 50 g |
| Assay Percent Range | 99.996% |
| Linear Formula | NH4VO3 |
| Fieser | 01,1057 |
| Merck Index | 15,565 |
| Solubility Information | Solubility in water: 5.1g/L (20°C). Other solubilities: insoluble in ethanol, 69.5g/L (96°C) |
| IUPAC Name | vanadiumoylol amine |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled. Toxic if swallowed. Causes serious eye irritation. Harmful if inhaled. Toxic to aquati |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. |
| EINECSNumber | 232-261-3 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.