Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUGT1 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB291399 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291399 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291399 Supplier Novus Biologicals Supplier No. NBP149707F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SUGT1 Polyclonal antibody specifically detects SUGT1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen SUGT1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias Protein 40-6-3, putative 40-6-3 protein, Sgt1, SGT1, suppressor of G2 allele of SKP1 (S. cerevisiae), SGT1B protein, suppressor of G2 allele of SKP1 homolog, suppressor of G2 allele of SKP1, S. cerevisiae, homolog of
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 315 and 365 of human Suppressor of G2 Allele of SKP1 using the numbering given in entry NP_001124384.1 (GeneID 10910).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10910
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.