Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUHW1 Antibody (OTI2H3), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP274409C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274409C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274409C Supplier Novus Biologicals Supplier No. NBP274409C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SUHW1 Monoclonal specifically detects SUHW1 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen SUHW1
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI2H3
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C19orf37, chromosome 19 open reading frame 37, Enzyme-like protein PIT13, MGC51082, Zfp428, zinc finger protein 428
Gene Symbols ZNF280A
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-333 of human SUHW1 (NP_542778) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 129025.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.