Learn More
Sulfanilic acid, ACS, 98.0-102.0%
Description
Sulfanilic acid is used as a reagent for chromatographic detection of histidine. It is also used as a chemical intermediate in organic synthesis. It is utilized in quantitative analysis of nitrate and nitrite ions. It plays an essential role in the manufacturing of azo dyes and in synthesis of sulfa drugs. It is used as a dopant for the chemical synthesis of polyaniline. It is used to prepare 2,4-diamino-6-p-sulphoanilinopyrimidine by reacting with 6-chloro-pyrimidine-2,4-diamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | ∼365°C |
| Quantity | 50 g |
| Beilstein | 908765 |
| Merck Index | 14,8926 |
| Solubility Information | Slightly soluble in water. Insoluble in ethanol,ether,caustic soda and sodium carbonate. |
| Formula Weight | 173.19 |
| Percent Purity | 98.0 to 102.0% |
| Grade | ACS |
| Physical Form | Powder |
| Chemical Name or Material | Sulfanilic acid |
Safety and Handling
| Hazard Category | H315-H317-H319 |
| Hazard Statement | GHS H Statement H315-H319-H317 Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. |
| Precautionary Statement | P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c |
| EINECSNumber | 204-482-5 |
| RTECSNumber | WP3895500 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.