Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfanilic acid sodium salt, MP Biomedicals™

Catalog No. p-4405251 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
50 g
250 g

Chemical Identifiers

CAS 6106-22-5
Molecular Formula C6H10NNaO5S
Molecular Weight (g/mol) 231.198
InChI Key MJFZIDUBUNIFLZ-UHFFFAOYSA-M
Synonym sodium sulfanilate dihydrate, unii-y31oen1192, 4-aminobenzenesulfonic acid, sodium salt dihydrate, sodium 4-aminobenzenesulfonate dihydrate, benzenesulfonic acid,4-amino-, sodium salt, hydrate 1:1:?, sulfanilic acid sodium salt dihydrate mi, acmc-1b9i8, c6h6nso3.na.2h2o, benzenesulfonic acid, 4-amino-, sodium salt, hydrate 1:1:2, 4h-1,4-benzoxazin-4-amine,2,3-dihydro hcl
PubChem CID 23687713
IUPAC Name sodium;4-aminobenzenesulfonate;dihydrate
SMILES C1=CC(=CC=C1N)S(=O)(=O)[O-].O.O.[Na+]

Specifications

Color White
Quantity 50 g
Formula Weight 195.2
Physical Form Powder
Chemical Name or Material Sulfanilic acid sodium salt
Recommended Storage Store at 2° to 8°C.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.