Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfatase-2/SULF2 Antibody (2B4) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP136727SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP136727SS 0.025 mL
NBP136727 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP136727SS Supplier Novus Biologicals Supplier No. NBP136727SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 6 publications

Sulfatase-2/SULF2 Monoclonal specifically detects Sulfatase-2/SULF2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready.

Specifications

Antigen Sulfatase-2/SULF2
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2B4
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry reported by customer review on FITC-conjugated antibody., ELISA 1:100-1:2000, Immunohistochemistry 1:250-1:500, Immunoprecipitation 1:10-1:100, Immunohistochemistry-Paraffin 1:250-1:500, CyTOF-ready
Gene Accession No. NP_940998
Gene Alias DKFZp313E091, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.14, extracellular sulfatase Sulf-2, FLJ44488, FLJ90554, hSulf-2, KIAA1247HSULF-2, MGC126411, SULF-2, sulfatase 2
Gene Symbols SULF2
Host Species Mouse
Immunogen Partial recombinant human Sulfatase 2 isoform b [Accession# NP_940998].
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 55959
Test Specificity Does not recognize human or mouse Sulfatase 1 or other family members.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.