Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfatase-2/SULF2 Antibody (2B4), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP136727IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP136727IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP136727IR Supplier Novus Biologicals Supplier No. NBP136727IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Sulfatase-2/SULF2 Monoclonal specifically detects Sulfatase-2/SULF2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Sulfatase-2/SULF2
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2B4
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Gene Alias DKFZp313E091, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.14, extracellular sulfatase Sulf-2, FLJ44488, FLJ90554, hSulf-2, KIAA1247HSULF-2, MGC126411, SULF-2, sulfatase 2
Gene Symbols SULF2
Host Species Mouse
Immunogen Partial recombinant human Sulfatase 2 isoform b [Accession# NP_940998].
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 55959
Test Specificity Does not recognize human or mouse Sulfatase 1 or other family members.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.