Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfatase-2/SULF2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB151774 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151774 50 μg
NB151773 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB151774 Supplier Novus Biologicals Supplier No. NBP31191050UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sulfatase-2/SULF2 Polyclonal antibody specifically detects Sulfatase-2/SULF2 in Human samples. It is validated for Western Blot

Specifications

Antigen Sulfatase-2/SULF2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.05 - 0.1 ug/ml
Formulation PBS
Gene Alias DKFZp313E091, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.14, extracellular sulfatase Sulf-2, FLJ44488, FLJ90554, hSulf-2, KIAA1247HSULF-2, MGC126411, SULF-2, sulfatase 2
Host Species Rabbit
Immunogen Synthetic peptide derived from the human Sulfatase-2/SULF2 protein. (Uniprot: Q8IWU5)
Purification Method Antigen Affinity-purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 55959
Target Species Human
Content And Storage Store at -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.