Description
- For use in learning about streak tests
- Sulfur leaves an unexpected white streak and distinct smell
- For use in classroom identification labs and exercises
- Correlates well to Next Generation Science Standards Evidence Statements
Specifications
Specifications
| Product Type | Mineral |
| For Use With (Application) | Lessons on identifying minerals in unconventional ways |
| Packaging | Clear labeled and barcoded bag |
| Quantity | 12/Pk. |
| Specimen Size | 2 to 3 cm |
| Specimen Type | Sulfur |