Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfur, 99.5+%, refined

Catalog No. AC201220025
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum

Catalog No. AC201220025

Supplier: Thermo Scientific Chemicals 201220025

Only null left
Add to Cart

CAS: 7704-34-9 | S | 32.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7704-34-9
Molecular Formula S
Molecular Weight (g/mol) 32.06
MDL Number MFCD00085316
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
Synonym polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
PubChem CID 5362487
ChEBI CHEBI:26833
IUPAC Name sulfur
SMILES [S]

Specifications

Boiling Point 445.0°C
Melting Point 114.0°C
Color Yellow
Physical Form Powder
Quantity 2.5 kg
Assay Percent Range 99.5% min
Flash Point 168°C
Fieser 01,1118; 03,273; 05,632; 06,556; 14,291; 15,297
Merck Index 15,9099
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in alcohol and ether, soluble in carbon disulfide, benzene, (about 2.4%, at 30°C, much more at higher temps.),toluene,, acetone (2.65% at 25°C), chloroform (1.5% at 18°C)
Formula Weight 32.05
Percent Purity ≥99.5%
Packaging Plastic bottle
Chemical Name or Material Sulfur
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and w
EINECSNumber 231-722-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.